CLS3544
Corning® 384 well microplate, low volume
NBS™ (non-binding surface), black polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates
About This Item
Recommended Products
material
black polystyrene
clear bottom
flat bottom
round clear wells
sterility
non-sterile
feature
lid: no
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3544
well working volume
5-40 μL
wells
384 wells , low volume
color
black
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
NBS™ (non-binding surface)
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | CLS3643 | CLS3821BC | CLS3540 |
---|---|---|---|
suitable for (molecular, validated for the FLIPR reader) | suitable for (molecular, validated for the FLIPR reader) | suitable for (molecular, validated for the FLIPR reader) | suitable for (molecular, validated for the FLIPR reader) |
non-sterile | sterile | non-sterile | non-sterile |
lid: no | lid, skirt | lid: no, skirt, with generic barcode | lid: no, skirt |
pack of 10 | pack of 10, case of 50 | pack of 10 | pack of 10, case of 50 |
Corning 3544 | Corning 3643 | Corning 3821BC | Corning 3540 |
General description
- Round well design with a clear flat bottom
- 2.8 mm distance between well bottoms to plate bottom
- 50μL total well volume, with recommended working volume of 5-40μL
- High base plate design is compatible with automation and for bar coding
- Validated by Molecular Devices for the FLIPR reader. Outer wells aligned for proper viewing with FLIPR
- Ideal for miniaturized 384 well fluorescent homogeneous assays
- Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service