Skip to Content
Merck
All Photos(1)

Key Documents

CLS3347

Corning® 384 Well Deep Well Plate

clear polypropylene, V-bottom, non-sterile, deep well, lid: no, pack of 10

Synonym(s):

384 deep well storage block, 384 well assay block, 384 well storage block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
V-bottom
clear bottom
clear polypropylene
polypropylene (plates, blocks and sealing mats)
square translucent wells
size:
384 wells
Pricing and availability is not currently available.

material

V-bottom
clear bottom
clear polypropylene
polypropylene (plates, blocks and sealing mats)
square translucent wells

sterility

(non-treated)
non-sterile

feature

lid: no
lid: no
skirt
deep well
plate format: 384 well storage block

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3347

size

384 wells

well volume

240 μL

color

clear

suitability

suitable for (general storage, liquid handling; automated applications)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Ideal for high throughput applications requiring added volume
  • Improved sample recovery through the specially designed wells that feature a square geometry that tapers to a V-bottom point
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Certified DNase- and RNase-free
Corning offers a complete range of polypropylene storage systems consisting of plates, blocks, and sealing mats. All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 384 well polypropylene plates and blocks are built to proposed industry standard footprint. All plates and blocks are automation-friendly and solvent resistant. The 384 well plates and blocks are certified DNase/RNase free and feature rounded bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service