CLS11656
Corning® closed system bottle
capacity 250 mL, polystyrene, male MPC connector, sterile
Synonym(s):
storage vessel
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
clear bottle
polystyrene
round bottle
wide-mouth bottle
sterility
sterile
feature
cap
male MPC connector
packaging
pkg of 6 ea
manufacturer/tradename
Corning 11656
capacity
250 mL
General description
Corning polystyrene Storage Bottles are now available as closed system solutions. They are sterile and arrive individually double-bagged and preassembled.
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
- Made from optically clear polystyrene
- Sterilized to SAL 10-6
Legal Information
Corning is a registered trademark of Corning, Inc.