Skip to Content
Merck
All Photos(1)

Key Documents

2-179-E11

from Mouse x Mouse Hybridoma, 92030602, Reactivity Anti-TNF alpha, human )

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

NS0

Reactivity

Anti-TNF alpha, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

Cell Line Origin

Mouse NS0 x Mouse BALB/c spleen

Cell Line Description

This strongly neutralising antibody is directed against natural and recombinant human TNF alpha. It does not cross-react with TNF ß. It cross-reacts with simian TNF alpha but not with TNF alpha or ß of other species. It can be paired in a sandwich ELISA with 357-101-4. Disclaimer required.

Immunogen

Highly purified recombinant human tumour necrosis factor alpha

Application

Immunoassays, TNF alpha neutralisation, immunohistochemistry

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS) (heat inactivated)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service