Skip to Content
Merck
All Photos(1)

Key Documents

Clone C6/36

89051705, insect larvae, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Clone C6/36, 89051705

biological source

insect larvae

packaging

tube of 5 μg 89051705-DNA-5UG
pkg of vial of cells 89051705-1VL

growth mode

Adherent

karyotype

Diploid

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mosquito larvae, Aedes albopictus

Cell Line Description

Derived from Aedes albopictus cells. Useful for propagation of Dengue virus and flaviviruses. Cells are non-anchorage dependant, not tumourigenic and maintain a diploid chromosome number.

Application

Clone C6/36 has been used to screen anti-Wolbachia compounds. It has also been used to study the effects of tick-borne encephalitis virus (TBEV) vaccine.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS) (Sodium Bicarbonate (NaHCO3) level of 1.1g/l is recommended).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 5000 - 30000 cells/cm2 . Cells are dislodged by mechanical agitation; 5% CO2; 28°C. Freeze in 10% glycerol.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service