1321N1 Cell Line human
86030402, human brain (astrocytoma), Glial
Synonym(s):
1321-N1 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
product name
1321N1 Cell Line human, from human brain(astrocytoma), 86030402
biological source
human brain (astrocytoma)
growth mode
Adherent
karyotype
Not specified
morphology
Glial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human brain astrocytoma
Cell Line Description
1321N1 is a human astrocytoma cell line isolated in 1972 as a sub clone of the cell line 1181N1 which in turn was isolated from the parent line U-118 MG (one of a number of cell lines derived from malignant gliomas by J Ponten .1321N1 has been shown to have very similar STR profile data to U-118 MG.
Application
Human 1321N1 Cell Line has been used to study how the multiscale morphology of functional thin films affects the in vitro behavior of human neural astrocytoma 1321N1 cells. It has also been used to study the expression and reactivity of Caytaxin protein.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 9,11
D16S539: 12,13
D5S818: 11
D7S820: 9
THO1: 6
TPOX: 8
vWA: 18
CSF1PO: 11,12
D13S317: 9,11
D16S539: 12,13
D5S818: 11
D7S820: 9
THO1: 6
TPOX: 8
vWA: 18
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service