Skip to Content
Merck
All Photos(1)

Key Documents

AXYSCT150SSO

Screw cap tubes and caps with O-rings, self-standing

size 1.5 mL, orange, pkg of 8x500caps/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc SCT-150-SS-O
Corning

packaging

pkg of 8x500caps/cs

size

1.5 mL

color

orange

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
AXYSCT200SSOAXYSCT050SSCAXYSCT050SSG
packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs

size

1.5 mL

size

2.0 mL

size

0.5 mL

size

0.5 mL

color

orange

color

orange

color

clear

color

green


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service