AXYPCR96HSC
PCR amplification plate
half skirt, 96 well, clear
Synonym(s):
96 multiwell plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
clear polypropylene wells
sterility
non-sterile
feature
skirt
packaging
case of 50 ea
manufacturer/tradename
Axygen Inc
Corning
technique(s)
PCR: suitable
size
96 wells
working volume
200 mL
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service