AXYPCR05V
PCR tubes with flat caps
thin wall, size 0.5 mL, pkg of 10x1000tubes/cs, violet
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Axygen Inc PCR-05-V
Corning
packaging
pkg of 10x1000tubes/cs
size
0.5 mL
color
violet
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service