Skip to Content
Merck
All Photos(1)

Key Documents

92416

Sigma-Aldrich

D-Mannitol 1-phosphate lithium salt

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15O9P · xLi+
CAS Number:
Molecular Weight:
262.15 (free acid basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

Assay

≥95% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)(O)O

InChI

1S/C6H15O9P/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14/h3-11H,1-2H2,(H2,12,13,14)/t3-,4-,5-,6-/m1/s1

InChI key

GACTWZZMVMUKNG-KVTDHHQDSA-N

Application

Mannitol-1-P can be used to detect the transphosphorylation activity of D-mannitol-specific enzyme II expreβed in spheroplasts.

Biochem/physiol Actions

D-Mannitol 1-phosphate is an intermediate in the fructose and mannose metabolism. It takes part in the phosphoenolpyruvate-dependent PEP group translocation (phosphotransferase system or PTS), a major mechanism used by bacteria for carbohydrate uptake.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Sutrina et al.
The Journal of biological chemistry, 262(6), 2636-2641 (1987-02-25)
The mannitol specific Enzyme II of the phosphoenolpyruvate: sugar phosphotransferase system of Escherichia coli catalyzes an exchange reaction in which a phosphoryl moiety is transferred from one molecule of the heat stable phosphocarrier protein HPr to another. An assay was
W J Wannet et al.
Antonie van Leeuwenhoek, 77(3), 215-222 (2004-06-11)
Acid phosphatase [AP; EC 3.1.3.2], a key enzyme involved in the synthesis of mannitol in Agaricus bisporus, was purified to homogeneity and characterized. The native enzyme appeared to be a high molecular weight type glycoprotein. It has a molecular weight
André B Canelas et al.
Biotechnology and bioengineering, 100(4), 734-743 (2008-04-03)
The coenzyme NAD plays a major role in metabolism as a key redox carrier and signaling molecule but current measurement techniques cannot distinguish between different compartment pools, between free and protein-bound forms and/or between NAD(H) and NADP(H). Local free NAD/NADH
H Rosenberg et al.
Journal of bacteriology, 158(1), 69-72 (1984-04-01)
A mutant (mtlD) strain of Escherichia coli unable to oxidize mannitol-1-phosphate to fructose-6-phosphate was used to study the fate of mannitol-1-phosphate. D-[1-14C]mannitol entered the cells via the phosphotransferase system and was phosphorylated equally at carbon 1 or 6. The label
The Intramembrane Topography of the Mannitol-specific Enzyme II of the Escherichia coli. Phosphotransferase System
Jacobson, G.
The Journal of Biological Chemistry, 285, 2955-2959 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service