TSS100
Total Suspended Solids 100 mg/L Calibration Standard
certified reference material
Synonym(s):
TSS Reference Solution (100 mg/L)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
CofA
certificate is enclosed in each package
manufacturer/tradename
RTC TSS100-100
concentration
in H2O
application(s)
environmental
format
single component solution
storage temp.
2-30°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Total Suspended Solids 100 mg/L Calibration Standard for R&D and Analytical use only.
Features and Benefits
- Packed in High Density Polyethylene (HDPE) bottle
- A certificate of analysis (CoA) is included with the product
- Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and International System of units (SI)
- Balance used to prepare the standard is calibrated to ISO 17025 requirements
- Stable solution with long minimum shelf life
Other Notes
Values of analytes vary lot to lot.
related product
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service