Skip to Content
Merck

PHR1877

Supelco

Valsartan Related Compound C

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine phenylmethyl ester, (N-VALERYL-N-{[2’-(1HTETRAZOLE-5-YL)BIPHENYL-4-YL]METHYL}-L-VALINEBENZYL ESTER), (S)-N-Valeryl-N-([2′-(1H-tetrazole-5-yl)-biphenyl-4-yl]methyl)valine benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N5O3
Molecular Weight:
525.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1708795

API family

valsartan

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 30 mg

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

SMILES string

CCCCC(=O)N(Cc1ccc(cc1)-c2ccccc2-c3nnn[nH]3)[C@@H](C(C)C)C(=O)OCc4ccccc4

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
W263508W234907W237701
Linalool ≥97%, FCC, FG

W263508

Linalool

organoleptic

green; melon; floral; sweet

organoleptic

lemon; orange; floral; sweet

organoleptic

musty; fruity; sweet

organoleptic

apple; grape; fruity; waxy; floral

biological source

synthetic

biological source

synthetic

biological source

synthetic

biological source

synthetic

food allergen

no known allergens

food allergen

no known allergens

food allergen

no known allergens

food allergen

no known allergens

agency

follows IFRA guidelines, meets purity specifications of JECFA

agency

follows IFRA guidelines, meets purity specifications of JECFA

agency

follows IFRA guidelines, meets purity specifications of JECFA

agency

follows IFRA guidelines, meets purity specifications of JECFA

assay

≥95%

assay

≥97%

assay

≥98%

assay

≥99%

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3176 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service