Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 4 vials x 5 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
A unit of SRM 633a comprises four vials, with each vial measuring 5 g of portland cement ground to pass through a 75 µm (No. 200) sieve, with each vial sealed in a foil pouch. For more information, please refer to the COA and SDS
SRM 633A_cert
SRM 633A_SDS
SRM 633A_cert
SRM 633A_SDS
Application
The Standard Reference Material (SRM) is primarily intended for use in evaluating methods used for the analysis of cements and materials with similar matrix.
Features and Benefits
- Certified mass fractions values and Reference mass fraction values for various constituents of SRM 633a are provided based on an as-received basis.
- Maintenance and expiration of SRM certification, instructions for handling, storage, and use are provided on NIST certificate.
Other Notes
Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminum Oxide (Al2O3), Ferric Oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulfur trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Zinc oxide (ZnO), Manganese(III) oxide (Mn2O3), Chlorine (Cl), Strontium oxide (SrO), Mercury (Hg)
Reference/Informational value is provided for the following analytes. Chromium(III) oxide (Cr2O3), Sulfide Sulfur, Calcium oxide (CaO), Fluoride (F–), Barium oxide (BaO)
See certificate for values and more details at nist.gov/SRM.
Silicon dioxide (SiO2), Aluminum Oxide (Al2O3), Ferric Oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulfur trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5), Zinc oxide (ZnO), Manganese(III) oxide (Mn2O3), Chlorine (Cl), Strontium oxide (SrO), Mercury (Hg)
Reference/Informational value is provided for the following analytes. Chromium(III) oxide (Cr2O3), Sulfide Sulfur, Calcium oxide (CaO), Fluoride (F–), Barium oxide (BaO)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service