NIST53E
Lead-base bearing metal (84Pb - 10Sb - 6Sn)
NIST® SRM® 53e
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 150 g
manufacturer/tradename
NIST®
technique(s)
X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable
format
matrix material
Related Categories
General description
SRM 53E_cert
SRM 53E_SDS
Other Notes
Antimony (Sb), Tin (Sn), Copper (Cu), Bismuth (Bi), Arsenic (As), Nickel (Ni), Iron (Fe)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1 Oral - STOT RE 2 - STOT RE 2 Inhalation
Target Organs
Central nervous system,Blood,Immune system,Kidney, Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service