NIST2570
Lead paint film for portable x-ray fluorescence analyzers
NIST® SRM® 2570, blank (color code: white)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 1 film
manufacturer/tradename
NIST®
technique(s)
XRF: suitable
application(s)
coatings
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This Standard Reference Material (SRM) is intended as a blank for the Lead Paint Film for Building Surfaces SRMs, which are used for validation of results from portable, hand-held, X-ray fluorescence analyzers when testing for lead in paint coatings on interior and exterior building surfaces. A unit of SRM 2570 consists of a white polyester sheet, approximately 7.6 cm wide, 10.2 cm long, and 0.2 mm thick, coated with a single, lacquer layer, approximately 0.04 mm thick. Each unit is over-coated with a clear, thin, plastic laminate to protect the surface from abrasion. For more information, please refer to the COA and SDS.
SRM 2570_cert
SRM 2570_SDS
SRM 2570_cert
SRM 2570_SDS
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lead (Pb)
See certificate for values and more details at nist.gov/SRM.
Lead (Pb)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service