M1470000
Methyl 2-(4-chlorophenoxy)-2-methylpropionate
European Pharmacopoeia (EP) Reference Standard
About This Item
grade
pharmaceutical primary standard
API family
clofibrate
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Clc1ccc(cc1)OC(C)(C)C(=O)OC
InChI
1S/C11H13ClO3/c1-11(2,10(13)14-3)15-9-6-4-8(12)5-7-9/h4-7H,1-3H3
InChI key
UXIVINXAZVEIMC-UHFFFAOYSA-N
General description
Application
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service