Skip to Content
Merck
All Photos(1)

Key Documents

BCR483

Sewage sludge amended soil (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Analysis Note

For more information please see:
BCR483

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Osyczka et al.
Ecotoxicology and environmental safety, 174, 26-34 (2019-03-01)
The epigeic lichens Cladonia rei and Diploschistes muscorum are effective heavy-metal-tolerant colonisers of highly polluted and disturbed sites. In this study we compare their bioaccumulation capacities, accumulation patterns, and responses to heavy-metal stress, as expressed in terms of cell membrane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service