Skip to Content
Merck
All Photos(1)

Key Documents

7620026000BC

Millipore

Gemini Sample Rack

BioControl Systems, for use with Gemini®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.84

Pricing and availability is not currently available.

packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems
Gemini®

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

application(s)

food and beverages

compatibility

for use with Gemini®

storage temp.

room temp

Legal Information

Gemini is a registered trademark of Stratec

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service