Skip to Content
Merck
All Photos(1)

Key Documents

76078

SAFC

L-Methionine sulfoximine

Pharma Manufacturing

Synonym(s):

L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
180.23
Beilstein:
6175446
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

Quality Level

Assay

≥96% (rel., HPLC)

form

solid

optical activity

[α]20/D +10.8 to +14.0°, c = 1 in H2O

concentration

95-102 % (w/w) (HPLC)

impurities

≤0.1% each single unknown impurity (rel.)
≤0.5% sulfated ash
≤0.5% water (Karl Fischer)
≤1% methionine sulfone (rel.)
≤1% methionine sulfoxide (rel.)
≤10 mg/kg azide
≤2% methionine (rel.)
≤5000 ppm ethanol (GC-HS)

mp

>210 °C (dec.) (lit.)

suitability

complies for H-NMR
complies for identity (HPLC)

SMILES string

CS(=N)(=O)CC[C@H](N)C(O)=O

InChI

1S/C5H12N2O3S/c1-11(7,10)3-2-4(6)5(8)9/h4,7H,2-3,6H2,1H3,(H,8,9)/t4-,11?/m0/s1

InChI key

SXTAYKAGBXMACB-DPVSGNNYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Wang et al.
PloS one, 6(3), e18233-e18233 (2011-04-13)
The membrane-proximal external region (MPER) of the HIV-1 gp41 consists of epitopes for the broadly cross-neutralizing monoclonal antibodies 2F5 and 4E10. However, antigens containing the linear sequence of these epitopes are unable to elicit potent and broad neutralizing antibody responses
Hsiu-Ni Kung et al.
PLoS genetics, 7(8), e1002229-e1002229 (2011-08-20)
Although significant variations in the metabolic profiles exist among different cells, little is understood in terms of genetic regulations of such cell type-specific metabolic phenotypes and nutrient requirements. While many cancer cells depend on exogenous glutamine for survival to justify
Evan T Brower et al.
Biochemistry, 48(4), 779-785 (2009-01-28)
The initial events of HIV-1 cell infection involve the sequential binding of the viral envelope glycoprotein gp120 to the cellular CD4 receptor and the coreceptor, usually CCR5 or CXCR4. Binding to the coreceptor triggers the chain of events that culminates
Marilynn A Larson et al.
The Biochemical journal, 462(2), 329-335 (2014-06-12)
Human BChE (butyrylcholinesterase) protects against the toxicity of organophosphorus nerve agents and pesticides. BChE purified from human plasma is limited and pathogen carry-over is a concern. Unlike the native BChE tetrameric complex with a residence time of days, rBChE (recombinant
Aneesh Vijayan et al.
PloS one, 10(7), e0133595-e0133595 (2015-07-25)
In the HIV vaccine field, there is a need to produce highly immunogenic forms of the Env protein with the capacity to trigger broad B and T-cell responses. Here, we report the generation and characterization of a chimeric HIV-1 gp120

Protocols

Separation of L-Methionine sulfone; L-Methionine sulfoximine

Separation of L-Methionine sulfone; L-Methionine sulfoximine; L-Methionine sulfoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service