51082
Bromhexine hydrochloride
analytical standard
Synonym(s):
2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride, Bromohexine hydrochloride
About This Item
grade
analytical standard
Quality Level
Assay
98.0-102.0% (AT)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
Cl.CN(Cc1cc(Br)cc(Br)c1N)C2CCCCC2
InChI
1S/C14H20Br2N2.ClH/c1-18(12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17;/h7-8,12H,2-6,9,17H2,1H3;1H
InChI key
UCDKONUHZNTQPY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Cough-cold syrup using high performance liquid chromatography (HPLC).
- Pharmaceutical formulations using gas liquid chromatography (GLC) and high performance liquid chromatography (HPLC).
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service