Skip to Content
Merck

472662

Sigma-Aldrich

Ethyl viologen diiodide

99%

Synonym(s):

1,1′-Diethyl-4,4′-bipyridinium diiodide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H18I2N2
CAS Number:
Molecular Weight:
468.12
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99%

form

powder

technique(s)

titration: suitable

mp

275 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

SMILES string

[I-].[I-].CC[n+]1ccc(cc1)-c2cc[n+](CC)cc2

InChI

1S/C14H18N2.2HI/c1-3-15-9-5-13(6-10-15)14-7-11-16(4-2)12-8-14;;/h5-12H,3-4H2,1-2H3;2*1H/q+2;;/p-2

InChI key

ZXBJDSANDCPCLD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl viologen diiodide serves as an electrolyte to conduct cyclic voltammetry to investigate the kinetic process of the redox targeting reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service