40952
Nickels and Leesment ChromoSelect Selective Supplement
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171614
NACRES:
NB.21
sterility
sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Lactococcus spp.
Leuconostoc spp.
Related Categories
Application
An antibiotic supplement used for the selective isolation of citrate-fermenting lactic acid bacteria.
Components
Composition:
(per vial, sufficient for 500 ml)
Vancomycin 100 mg
(per vial, sufficient for 500 ml)
Vancomycin 100 mg
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service