Skip to Content
Merck
All Photos(1)

Key Documents

36543

Supelco

2,4,6-Trichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
776729
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor pressure

1 mmHg ( 76.5 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

246 °C (lit.)

mp

64-66 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Trichlorophenol belongs to the class of chlorophenols, which occur as industrial wastes and direct environmental pollutants.

Application

2,4,6-Trichlorophenol may be used as an analytical reference standard for the determination of the analyte in environmental samples using gas chromatography coupled to mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jijun Gao et al.
Chemosphere, 71(6), 1181-1187 (2007-11-27)
The chlorophenol pollutants (CPs) have been reported to occur at relatively high concentrations in some Chinese waters. To map the distribution of CPs in the surface water throughout China, samples were collected from over 600 sites in the seven major
Meiqin Hu et al.
Environmental science & technology, 46(16), 9005-9011 (2012-07-26)
Molecular iodine has been studied, for the first time, as a sensitizer for the degradation of 2,4,6-trichlorophenol (TCP) in aqueous solution under visible light (λ ≥ 450 nm). TCP was degraded in the presence of commercial I(2), but the reaction
Stefan Franzen et al.
The journal of physical chemistry. B, 116(5), 1666-1676 (2011-12-14)
Kinetic and structural studies have shown that peroxidases are capable of the oxidation of 2,4,6-trichlorophenol (2,4,6-TCP) to 2,6-dichloro-1,4-benzoquinone (2,6-DCQ). Further reactions of 2,6-DCQ in the presence of H(2)O(2) and OH(-) yield 2,6-dichloro-3-hydroxy-1,4-benzoquinone (2,6-DCQOH). The reactions of 2,6-DCQ have been monitored
Nadavala Siva Kumar et al.
Colloids and surfaces. B, Biointerfaces, 94, 125-132 (2012-03-01)
Biosorption of 2,4,6-trichlorophenol (2,4,6-TCP) from aqueous solution by biomass prepared from Acacia leucocephala bark, an agricultural solid waste has been investigated in the present study. All the experiments are carried out by batch mode technique. The resulting biosorbent was characterized
Supreet S Bahga et al.
Electrophoresis, 33(6), 1048-1059 (2012-04-25)
We present a novel method of creating concentration cascade of leading electrolyte (LE) in isotachophoresis (ITP) by using bidirectional ITP. ITP establishes ion-concentration shock waves between high-mobility LE and low-mobility trailing electrolyte (TE) ions. In bidirectional ITP, we set up

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service