Skip to Content
Merck
All Photos(1)

Key Documents

34139

Supelco

Fumonisin B1 solution

~50 μg/mL in acetonitrile: water, analytical standard

Synonym(s):

FB1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H59NO15
CAS Number:
Molecular Weight:
721.83
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile: water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H](N)[C@@H](O)C[C@H](O)CCCC[C@@H](O)C[C@H](C)C[C@H](OC(C[C@H](C(O)=O)CC(O)=O)=O)[C@H](OC(C[C@H](C(O)=O)CC(O)=O)=O)[C@H](C)CCCC

InChI

1S/C34H59NO15/c1-5-6-9-20(3)32(50-31(44)17-23(34(47)48)15-29(41)42)27(49-30(43)16-22(33(45)46)14-28(39)40)13-19(2)12-24(36)10-7-8-11-25(37)18-26(38)21(4)35/h19-27,32,36-38H,5-18,35H2,1-4H3,(H,39,40)(H,41,42)(H,45,46)(H,47,48)/t19-,20+,21-,22+,23+,24+,25+,26-,27-,32+/m0/s1

InChI key

UVBUBMSSQKOIBE-DSLOAKGESA-N

General description

Certan Vial
Fumonisin B1 belongs to the family of toxins known as fumonisins. It is produced by several species of Fusarium molds, primarily by Fusarium verticillioide. It is one of the major mycotoxin contaminant in corn and corn-based foods. Furthermore it is hepatotoxic and nephrotoxic.

Application

Fumonisin B1 maybe used in the fumonisin injection experiment to study pulmonary edema and hydrothorax in swine produced by fumonisin B1.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aspergillus: Molecular Biology and Genomics.
Machida M and Gomi K.
Science, 146-147 (2010)
Pulmonary edema and hydrothorax in swine produced by fumonisin B1, a toxic metabolite of Fusarium moniliforme.
Harrison BM, et al.
Journal of Veterinary Diagnostic Investigation : Official Publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 217-221 (1990)
Agriculture and Food Production.
Khachatourians GG and Arora DK.
Science, 181-182 (2002)
Arya Bagus Boedi Iswanto et al.
Plant physiology, 184(1), 407-420 (2020-07-09)
Plasma membranes encapsulated in the symplasmic nanochannels of plasmodesmata (PD) contain abundant lipid rafts, which are enriched with sphingolipids (SLs) and sterols. Reduction of sterols has highlighted the role played by lipid raft integrity in the intercellular trafficking of glycosylphosphatidylinositol
Ronald T Riley et al.
Molecular nutrition & food research, 56(9), 1445-1455 (2012-07-21)
Fumonisins (FB) are mycotoxins found in maize. The purpose of this study was to (i) determine the relationship between FB(1) , FB(2) , and FB(3) intake and urinary excretion in humans, (ii) validate a method to isolate urinary FB on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service