Skip to Content
Merck
All Photos(2)

Key Documents

275980

Sigma-Aldrich

3-Methoxydiphenylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NHC6H5
CAS Number:
Molecular Weight:
199.25
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

98%

form

powder or crystals

mp

72-74 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1cccc(Nc2ccccc2)c1

InChI

1S/C13H13NO/c1-15-13-9-5-8-12(10-13)14-11-6-3-2-4-7-11/h2-10,14H,1H3

InChI key

MKASXAGBWHIGCF-UHFFFAOYSA-N

Application

3-Methoxydiphenylamine can be used in the preparation of 3- methoxydiphenylamine-4-diazonium salts (MDP-4-DS) and the corresponding diazoresins. Diazonium salts and diazoresins find applications in dyes, stains, indicators, general laboratory use, and industrial applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of Epoxy-Based Polymeric Azo Dyes
R-q SUN, et al.
Journal of Functional Polymers / Gong Neng Gao Fen Zi Xue Bao, 3 (2008)
Synthesis and characterization of diphenylamine diazonium salts and diazoresins
Zhao C, et al.
Angew. Makromol. Chem., 259(1), 77-82 (1998)
Sirivipa Mongkolkitikul et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 112, 20-27 (2017-11-08)
The pristine pectin hydrogels and conductive polymer/hydrogel blends were prepared by the solution casting using ferrous chloride (FeCl2) and citric acid as the crosslinking agents, and ibuprofen as the model drug and the doping agent. A conductive polymer, poly(3-methoxydiphenylamine), was
Dongkun Yu et al.
Chemistry, an Asian journal, 14(19), 3350-3356 (2019-08-27)
Eutectic molecular liquids (EMLs) based on hydrogen-bonding interaction and π-π stacking were prepared. We found that the thermodynamic properties like initial decomposition temperature and glass transition temperature of EMLs are mainly dominated by the hydrogen bond donor, which is beneficial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service