Key Documents
26294
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1005SLPT (for Purge & Trap), volume 5 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass body
needle size
(not included)
feature
This syringe allows you to easily remove the syringe plunger and load water samples into the syringe barrel
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81570
volume
5 mL
General description
Purge and Trap Syringes
This purge and trap version of the SampleLock syringe is designed for analyzing drinking water samples according US EPA purge and trap concentration techniques (EPA methods 502.1, 502.2, 503.1, 524.1, and 524.2).
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Customers Also Viewed
Slide 1 of 1