18021
Sulfur Standard for ICP
TraceCERT®, 1 g/L S in H2O (nominal concentration)
Synonym(s):
Sulfur ICP Standard Solution
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
concentration
1 g/L S in H2O (nominal concentration)
technique(s)
ICP: suitable
application(s)
industrial qc
pharmaceutical
format
single component solution
1 of 4
This Item | GF38272507 | GF18443369 | GF02051491 |
---|---|---|---|
foil (as rolled) | foil (as rolled) | foil (as rolled) | foil (as rolled) |
light tight (lt) | light tight (lt) | light tight (lt) | light tight (lt) |
150 mm × 150 mm × 0.025 mm | 25 mm × 25 mm × 0.025 mm | 25 mm × 25 mm × 0.0125 mm | 100 mm × 100 mm × 0.01 mm |
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
Analysis Note
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service