SIPK0SIA1
SimpliPak® Purification Cartridge
For Simplicity® water purification systems with Elix® / RO / distilled water feed
Synonym(s):
SimpliPak® 1 Purification Cartridge
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
feature
can remove trace contaminants
packaging
pkg of 1 unit
manufacturer/tradename
SimpliPak®
resistivity
>18.0 MΩ-cm (at 25 °C)
pore size
0.05 μm pore size
compatibility
for use with Simplicity® UV
for use with Simplicity®
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
- SimpliPak® 1 Purification Cartridge is designed to remove ionic and organic contaminates down to trace levels when fed with Elix® / RO / distilled water feed.
- The cartridge is constructed with clean and high-purity materials to minimize the leaching of extractable contaminants.
Application
Removes ionic and organic contaminants down to trace levels.
Features and Benefits
- Genuine consumable for the replacement of SIPK0SIA1 - SimpliPak® 1 Purification Cartridge.
- Care-free maintenance: easily installed and replaced.
- Generates ionic and organic contaminant-free water by ion exchange, and activated carbon technologies.
- Certificate of Quality available online.
Legal Information
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service