MMDRNTUBE
MILLIFLEX OASIS® drain tubing
for use with Milliflex Oasis® systems
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.51
material
silicone
packaging
pack of 1 unit
manufacturer/tradename
Milliflex Oasis®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis® systems
shipped in
ambient
Related Categories
General description
MILLIFLEX OASIS® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The MILLIFLEX OASIS® pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. The MILLIFLEX OASIS® drain tubing is used to discard the liquid waste during the usage of the pump.
Application
MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical Products Analysis.
Legal Information
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service