Skip to Content
Merck
All Photos(2)

Key Documents

MABS1183

Sigma-Aldrich

Anti-Periostin Antibody, clone 5B2.3

clone 5B2.3, from mouse

Synonym(s):

Periostin, OSF-2, Osteoblast-specific factor 2, PN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B2.3, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... POSTN(10631)

General description

Periostin (UniProt Q15063; also known as OSF-2, Osteoblast-specific factor 2, PN) is encoded by the POSTN (also known as PSF2) gene (Gene ID 10631) in human. Periostin is an extracellular matrix (ECM) protein predominantly expressed in collagen-rich fibrous connective tissues, including heart valves, tendons, perichondrium, periosteum, and periodontal ligament (PDL). Periostin plays an essential role in the process of remodeling during tissue/organ development/repair and inflammation. Periostin derived from fibroblasts is shown to induce keratinocytes differentiation and proliferation. Periostin is a mediator for epidermal hyperplasia and is highly expressed in the dermis of both atopic dermatitis (AD) and psoriasis patients. Periostin level is also reported to be upregulated in patients with acute or chronic hepatitis and correlates well with serum TGF- 1 and TGF- 2 levels. The PDL-POSTN spliced isoform is predominantly expressed in PDL, it binds integrin v 3 with a higher affinity and induces strong activation of the integrin v 3-focal adhesion kinase (FAK) signaling pathway. Periostin is produced with a signal peptide (a.a. 1-21), the removal of which yields the mature protein with an N-terminal cysteine-rich EMI (a.a. 40-94) domain and four asciclin I or FAS1 domains (a.a. 97-230, 234-365, 368-492, 496-628). Alternative splicing generate several C-terminal domain variants with tissue-specific expression patterns.

Specificity

Clone 5B2.3 epitope lies within a region C-terminal to the FAS1 domains. Immunogen covered region is 100% present in isoforms 1 (OSF-2OS), 2 (OSF-2p1), and 5, but partially missing in spliced isoforms 3, 4 (PDL-POSTN), 6, and 7. Clone 5B2.3 immunoreactivity toward these latter isoforms is possible, but has not been confirmed.

Immunogen

GST-tagged recombinant human periostin C-terminal fragment.

Application

Detect Periostin using this mouse monoclonal Anti-Periostin, clone 5B2.3, Cat. No. MABS1183, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected periostin in human prostate cancer, breast and small intestine tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human stomach tissue lysate.

Western Blotting Analysis: 0.5 Āµg/mL of this antibody detected periostin in 10 Āµg of human stomach tissue lysate.

Target description

~78-93 kDa observed. The broad band appearance is consistent with the detetion of differentially glycosylated species of one or more spliced isoforms. 90.96/87.021/84.67/81.50/88.07/81.17/78.00 kDa (mature isoform 1/2/3/4/5/6/7) and 93.31/87.02/87.25/83.85/90.42/83.53/80.36 kDa (isoform 1/2/3/4/5/6/7 pro-form) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8Ā°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product? Ā 

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service