Skip to Content
Merck
All Photos(1)

Key Documents

MABF1988

Sigma-Aldrich

Anti-CMTM6 Antibody, clone RCT6

Synonym(s):

CKLF-like MARVEL transmembrane domain-containing protein 6;Chemokine-like factor superfamily member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

RCT6, monoclonal

mol wt

calculated mol wt 20.42 kDa
observed mol wt ~20 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CMTM6(54918)

Related Categories

General description

CKLF-like MARVEL transmembrane domain-containing protein 6 (UniProt: Q9NX76; also known as Chemokine-like factor superfamily member 6, CMTM6) is encoded by the CMTM6 gene (Gene ID: 54918) in human. CMTM6 is a ubiquitously expressed, multi-pass membrane protein of the family of MARVEL domain-containing proteins with 3 cytoplasmic domains, 4 transmembrane domains, and 2 extracellular domains. Its MARVEL domain is localized in amino acids 33-160. It serves as a master regulator of recycling and plasma membrane expression of PD-L1. It binds to PD-L1 and maintains its cell surface expression. Although it is not required for PD-L1 maturation, but it co-localizes with PD-L1 at the plasma membrane and in recycling endosomes, where it prevents PD-L1 from being targeted for lysosome-mediated degradation by preventing its STUB-1 mediated ubiquitination. It also stabilizes PD-L1 expression on antigen presenting cells and potentiates inhibitory signaling by PD-1 (CD279), ultimately triggering T-cell anergy. It enhances the ability of PD-L1-expressing tumor cells to inhibit T cell activity. (Ref.: Burr, ML., et al. (2017). Nature. 549(7670); 101-105; Mezzadra, R., et al. (2017). Nature. 549(7670); 106-110).

Specificity

Clone RCT6 is a mouse monoclonal antibody that detects human CMTM6. It targets an epitope within the C-terminal region.

Immunogen

A linear peptide corresponding to a sequence from the C-terminal region of human CKLF-like MARVEL transmembrane domain-containing protein 6 (CMTM6).

Application

Quality Control Testing

Evaluated by Western blotting in lysates from 8505c cells overexpressing CMTM6.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected CMTM6 in lysates from 8505C cells overexpressing CMTM6, but not in knockout or untransfected cells.

Tested Applications


Immunohistochemistry Applications: A representative lot detected CMTM6 in Immunohistochemistry applications (Mezzadra, R., et al. (2017). Nature. 549(7670):106-110).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CMTM6, clone RCT6, Cat. No. MABF1988, is a mouse monoclonal antibody that detects CMTM6 and is tested for use in Immunohistochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service