FCABS326PE
Milli-Mark® Anti-acetyl-Histone H4 Antibody-PE
Milli-Mark®, from rabbit
Synonym(s):
Histone H4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
conjugate
PE
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Milli-Mark®
technique(s)
flow cytometry: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... H4C1(8359)
Related Categories
General description
Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).
Specificity
Recognizes acetylated histone H4. Cross-reacts with acetylated histone H2B from Tetrahymena and weakly cross-reacts with acetylated histone H2B from HeLa cells. May crossreact with other acetylated proteins.
Immunogen
Tetrahymena histone H4 acetylated on lysines 5, 8, 12, and 16.
Application
Milli-Mark Anti-acetyl-Histone H4 Antibody-PE is a highly specific rabbit polyclonal antibody, that targets Histone H4 & has been tested in Flow Cytometry.
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Histones
Histone Modifying Proteins
Histones
Quality
Evaluated by flow cytometry using HeLa cells.
Target description
11 kDa Calculated
Physical form
Protein A purified
Purified rabbit polyclonal IgG conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.
Storage and Stability
Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.
Analysis Note
Control
HeLa cells
HeLa cells
Legal Information
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service