AP500A
Goat Anti-Mouse IgM µ chain Antibody, Alkaline Phosphatase conjugate, Species Adsorbed
Chemicon®, from goat
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Recommended Products
biological source
goat
Quality Level
conjugate
alkaline phosphatase conjugate
antibody form
affinity purified immunoglobulin
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
shipped in
wet ice
target post-translational modification
unmodified
Related Categories
General description
IgM constitutes about 10% of serum immunoglobulins. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as ′natural′ blood group antibodies. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. The gene for the mu constant region contains four domains separated by short intervening sequences.
Specificity
Reacts with heavy chain of mouse IgM. Absorbed for mouse IgG1, IgG2a, IgG2b, IgG3 and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins. Reactivity with other species has not been determined.
Application
Goat anti-Mouse IgM µ chain Antibody, Alkaline Phosphatase conjugate, Species Adsorbed detects level of Mouse IgM µ chain & has been published & validated for use in ELISA.
Physical form
Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service