Skip to Content
Merck
All Photos(2)

Key Documents

ABF118

Sigma-Aldrich

Anti-FPR2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

N-formyl peptide receptor 2, FMLP-related receptor I, FMLP-R-I, Formyl peptide receptor-like 1, HM63, Lipoxin A4 receptor, LXA4 receptor, RFP, FPR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FPR2(2358)

Related Categories

General description

Formyl peptide receptors (FPRs) are G protein coupled receptors critical for our immune response and the cellular activation and or suppression of immune cell activities. N-formyl peptides are produced by the degradation of either bacteria or host cells and serve as chemoattractant agents for white blood cells. There are three known formyl peptide receptors, FPR1, FPR2, and FPR3 all appear to have similar functions. Binding of N-formyl peptides activates neutrophils via the phosphatidylinositol-calcium second messenger system. In some cases binding leads to a down regulation of immune signals such as in the down regulation observed against the proinflammatory signaling agent leukotriene B4. Interestingly, FPRs also have endogenous ligands which appear to function in a developmental fashion and not directly as an immune function. For instance, FPR2’s endogenous ligand is a peptide named Cathelin-related antimicrobial peptide (CRAMP) which is critical for proper dendritic cell maturation. FRP2 is abundantly expressed in the lung, spleen and testis neutrophils. Because of their role in the immune cell activation, FPRs are also being investigated as possible targets for modulating immune caused diseases such as arthritis.

Immunogen

Epitope: extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human FPR2.

Application

Anti-FPR2 Antibody | ABF118 is an antibody against FPR2 Antibody for use in Western Blotting, Flow Cytometry.
Flow Cytometry Analysis: 1.0 µg of this antibody detected FPR2 in HL-60 cells.
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected FPR2 in 10 µg of HL-60 cell lysate.

Target description

~45 kDa observed. The calculated molecular weight of this protein is 39 kDa, but can be observed at ~45 kDa due to high glycosylation.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rima Chattopadhyay et al.
Free radical biology & medicine, 117, 119-131 (2018-02-07)
In recent years, various studies have demonstrated a role for endogenously derived specialized proresolving mediators such as resolvins in the resolution of inflammation. In exploring the signaling mechanisms, in the present study we show that Resolvin D1 (RvD1) reduces LPS-induced
Rima Chattopadhyay et al.
Redox biology, 12, 438-455 (2017-03-21)
Resolvins are a novel class of lipid mediators that play an important role in the resolution of inflammation, although the underlying mechanisms are not very clear. To explore the anti-inflammatory mechanisms of resolvins, we have studied the effects of resolvin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service