Skip to Content
Merck
All Photos(2)

Key Documents

AB5935

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody, phosphoSer 40

Chemicon®, from rabbit

Synonym(s):

Tyrosine Monooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

avian, rat, mouse

species reactivity (predicted by homology)

quail (100% sequence homology), nonhuman primates (100% sequence homology), human (100% sequence homology), porcine (100% sequence homology)

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer40)

Gene Information

human ... TH(7054)
rat ... Th(25085)

General description

Tyrosine hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing.

Specificity

Other species have not been tested.
Tyrosine Hydroxylase, phosphoSer40. The antibody recognizes a protein of 60 kDa corresponding to TH phosphorylated at Ser40. Depending on brain region being studied, protein loads and detection methods used some higher molecular weight bands may be detected.

Immunogen

Epitope: phospho-Ser40
Synthetic peptide from the phosphoSer40 of rat Tyrosine Hydroxylase.

Application

Anti-Tyrosine Hydroxylase Antibody, phosphoSer 40 detects level of Tyrosine Hydroxylase & has been published & validated for use in IC, IH & WB.
Immunohistochemistry:
A 1:1,000 dilution of a previous lot was used in IH.

Immunocytochemistry:
A 1:1,000 dilution of a previous lot was used in IC.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected TYROSINE HYDROX PSER40 on 10 μg of PC12 lysates.

Target description

60 kDa

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal liquid in buffer containing 10mM Hepes, pH 7.5, 150mM NaCl, 100 µg/mL BSA, and 50% glycerol.

Storage and Stability

Stable for 6 months at -20°C in undiluted from date of receipt.

Analysis Note

Control
POSITIVE CONTROL: brain (sympathetic nerve terminals) and adrenal glands.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis M Colon-Perez et al.
Neuropharmacology, 137, 178-193 (2018-05-08)
Among cathinone drugs known as bath salts, methylenedioxypyrovalerone (MDPV) exerts its potent actions via the dopamine (DA) system, and at intoxicating doses may produce adverse behavioral effects. Previous work by our group suggests that prolonged alterations in correlated neural activity
Zhenqiang Zhao et al.
Frontiers in cellular neuroscience, 10, 291-291 (2017-01-10)
Mouse embryonic fibroblasts (MEFs) and human foreskin fibroblasts (HFFs) are used for the culture of human embryonic stem cells (hESCs). MEFs and HFFs differed in their capacity to support the proliferation and pluripotency of hESCs and could affect cardiac differentiation
Tsung-Jung Ho et al.
Evidence-based complementary and alternative medicine : eCAM, 2017, 5642708-5642708 (2017-04-13)
Methamphetamine (METH) is a major drug of abuse worldwide, and no efficient therapeutic strategies for treating METH addiction are currently available. Continuous METH use can cause behavioral upregulation or psychosis. The dopaminergic pathways, particularly the neural circuitry from the ventral
Cristina Núñez et al.
Endocrinology, 150(7), 3118-3127 (2009-01-31)
Chronic opiate exposure induces neurochemical adaptations in the noradrenergic system. Enhanced responsiveness of the hypothalamo-pituitary-adrenal axis after morphine withdrawal has been associated with hyperactivity of ascending noradrenergic input from the nucleus of the solitary tract (NTS-A(2)) cell group to the
Gregory V Carr et al.
Frontiers in pharmacology, 8, 666-666 (2017-10-24)
Pharmacological studies indicate that dopamine D1-like receptors (D1 and D5) are critically involved in cognitive function. However, the lack of pharmacological ligands selective for either the D1 or D5 receptors has made it difficult to determine the unique contributions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service