8.51001
FMOC-RINK LINKER
p-[(R,S)-α-[1-(9H-Fluoren-9-yl)-methoxyformamido]- 2,4-dimethoxybenzyl]- phenoxyacetic acid Novabiochem®
Synonym(s):
FMOC-RINK LINKER, p-{(R,S)-α-[1-(9H-Fluoren-9-yl)-methoxyformamido]-2,4-dimethoxybenzyl}-phenoxyacetic acid
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
Assay
≥98% (TLC)
≥98.0% (HPLC)
form
powder
reaction suitability
reagent type: linker
manufacturer/tradename
Novabiochem®
mp
170-185 °C
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
SMILES string
N(C(c5c(cc(cc5)OC)OC)c4ccc(cc4)OCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3
InChI
1S/C32H29NO7/c1-37-22-15-16-27(29(17-22)38-2)31(20-11-13-21(14-12-20)39-19-30(34)35)33-32(36)40-18-28-25-9-5-3-7-23(25)24-8-4-6-10-26(24)28/h3-17,28,31H,18-19H2,1-2H3,(H,33,36)(H,34,35)
InChI key
UPMGJEMWPQOACJ-UHFFFAOYSA-N
Related Categories
General description
Literature references
[1] M. S. Bernatowicz, et al. (1989) Tetrahedron Lett., 30, 4645.
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service