Skip to Content
Merck

561306

P-Selectin, Human, Recombinant, CHO Cells

Synonym(s):

CD62P, GMP-140, LECAM-3, PADGEM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352202
Form:
lyophilized
Assay:
≥95% (SDS-PAGE)
Recombinant:
expressed in CHO cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in CHO cells

Quality Segment

assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.5 EU/μg Endotoxins (EU/μg of P-Selectin)

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human P-selectin consisting of 730 amino acids minus the transmembrane and cytoplasmic domains expressed in CHO cells. This glycoprotein is expressed on activated platelets and endothelial cells. A specific interaction between P-selectin and P-selectin glycoprotein ligand-1, present on numerous cell types, promotes adhesion and acts along with L-selectin in establishing an initial interaction between circulating leukocytes and the endothelium.

Biochem/physiol Actions

Shown to bind to P-selectin ligands on U937 cells.

Physical form

Lyophilized from Dulbecco’s D-PBS (containing calcium and magnesium), 1% sorbitol.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Reconstitute at 1 mg/ml in sterile H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library