Skip to Content
Merck
All Photos(1)

Key Documents

444288

Sigma-Aldrich

MMP-2 Inhibitor III

The MMP-2 Inhibitor III, also referenced under CAS 704888-90-4, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2 Inhibitor III, (2-((Isopropoxy)-(1,1ʹ-biphenyl-4-ylsulfonyl)-amino))-N-hydroxyacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O5S
CAS Number:
Molecular Weight:
364.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

methanol: 100 mg/mL
DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N(OC(C)C)CC(=O)NO)c1ccc(cc1)c2ccccc2

InChI

1S/C17H20N2O5S/c1-13(2)24-19(12-17(20)18-21)25(22,23)16-10-8-15(9-11-16)14-6-4-3-5-7-14/h3-11,13,21H,12H2,1-2H3,(H,18,20)

InChI key

PHGLPDURIUEELR-UHFFFAOYSA-N

General description

A cell-permeable biphenylsulfonamido-hydroxamate compound that acts a potent and Zn2+-binding site-targeting inhibitor of MMP-2 (IC50 = 12 nM). It exhibits good selectivity over MMP-9 and MMP-3 (IC50 = 0.2 and 4.5 µM, respectively) and shows practically no effect towards MMP-1 and MMP-7 (IC50 >50 µM). Shown to effectively suppress the invasiveness of HT1080 sarcoma cells grown on matrigel.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-2
Product does not compete with ATP.
Reversible: no
Target IC50: 12 nM against MMP-2

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Tuccinardi, T., et al. 2006. Bioorg. Med. Chem.14, 4260.
Rossello, A., et al. 2004. Bioorg. Med. Chem.12, 2441.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivian C Chioma et al.
Biological psychiatry, 89(10), 947-958 (2021-02-14)
Seeking addictive drugs is regulated by synaptic plasticity in the nucleus accumbens core and involves distinct plasticity in D1 and D2 receptor-expressing medium spiny neurons (D1/2-MSNs). However, it is unknown how differential plasticity between the two cell types is coordinated.
Yusuke Sakamaki et al.
Nephron. Experimental nephrology, 115(2), e22-e32 (2010-04-22)
The role of matrix metalloproteinases (MMPs) in the pathogenesis of glomerular injury appears to be complex. To investigate the role of individual MMPs, we examined the course of Adriamycin-induced albuminuria and glomerulosclerosis in mice lacking either a gelatinase (MMP-9) or

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service