Skip to Content
Merck
All Photos(1)

Key Documents

428205-M

Millipore

Lck Inhibitor

The Lck Inhibitor, also referenced under CAS 213743-31-8, controls the biological activity of Lck. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Lck Inhibitor, 4-Amino-5-(4-phenoxyphenyl)-7H-pyrrolo[3,2-d]pyrimidin-7-yl-cyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22N4O
CAS Number:
Molecular Weight:
370.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 17 mg/mL

storage temp.

2-8°C

SMILES string

[n]2(c3ncnc(c3c(c2)c4ccc(cc4)Oc5ccccc5)N)C1CCCC1

InChI

1S/C23H22N4O/c24-22-21-20(14-27(17-6-4-5-7-17)23(21)26-15-25-22)16-10-12-19(13-11-16)28-18-8-2-1-3-9-18/h1-3,8-15,17H,4-7H2,(H2,24,25,26)

InChI key

FMETVQKSDIOGPX-UHFFFAOYSA-N

General description

A cell-permeable pyrrolopyrimidine compound that acts as a potent, reversible, selective, and ATP-competitive inhibitor of Lck (IC50 at 5 µM ATP = <1 nM, 2 nM, 70 nM, 1.57 µM and 1.98 µM for lck64-509 Y394, Lckcd pY394, Src, Kdr and Tie-2, respectively; IC50 at 1 mM ATP = 16 µM, 66 nM, 126 nM, 420 nM and 5.18 µM for Lck64-509 Y394, Blk, Fyn, Lyn and Csk, respectively). Only minimally affects the activities of other kinases (IC50 = 3.2 µM, >33 µM, >50 µM and >50 µM for EGFR, PKC, CDC2/B and ZAP-70, respectively). Also shown to potently block T-cell receptor-stimulated IL-2 production in vitro (IC50<1-40 nM in Jurkat T cells) and in vivo (ED50 = 4 mg/kg in mice, ip.)

Biochem/physiol Actions

Primary Target
Lck₆₄₋₅₀₉ Y³⁹⁴
Target IC50: At5 µM ATP = <1 nM, 2 nM, 70 nM, 1.57 µM and 1.98 µM for lck64-509 Y394, Lckcd pY394, Src, Kdr and Tie-2, respectively; IC50 at 1 mM ATP = 16 µM, 66 nM, 126 nM, 420 nM and 5.18 µM for Lck64-509 Y394, Blk, Fyn, Lyn and Csk

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Burchat, A.F., et al. 2000. Bioorg. Med. Chem. Lett.10, 2171.
Arnold. L.D., et al. 2000. Bioorg. Med. Chem. Lett.10, 2167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service