345852
Geranylgeranyltransferase I, GST-Fusion, His•Tag®, Rat, Recombinant, E. coli
Geranylgeranyltransferase I, GST-Fusion, His•Tag, Rat, Recombinant, E. coli, contains a 74 kDa α- & 43 kDa β-subunit. The α-subunit is expressed as a GST-fusion construct with an N-terminal His•Tag.
Synonym(s):
GGTase I
About This Item
Recommended Products
recombinant
expressed in E. coli
Quality Level
Assay
≥90% (SDS-PAGE)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Biochem/physiol Actions
Warning
Physical form
Reconstitution
Other Notes
Zhang, F.L., et al. 1994. J. Biol. Chem.269, 23465.
Moomaw, J.F., and Casey, P.J., 1992. J. Biol. Chem.267, 17438.
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service