Skip to Content
Merck

345754

Ganglioside GQ1b, Tetrasialo, Tetraammonium Salt, Bovine Brain

≥98% (TLC), lyophilized, IgG, IgM, IgA spontaneous production enhancer, Calbiochem®

Synonym(s):

Ganglioside GQ1b, Tetrasialo, Tetraammonium Salt, Bovine Brain, IV3(Neu5Ac)2II3(Neu5Ac)2-GgOse4, 4NH₄

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C106H178N6O55 · 4NH4
Molecular Weight:
2488.71
UNSPSC Code:
12352211
NACRES:
NA.77
Assay:
≥98% (TLC)
Form:
lyophilized
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ganglioside GQ1b, Tetrasialo, Tetraammonium Salt, Bovine Brain, Ganglioside GQ1b enhances spontaneous IgG, IgM, and IgA production in human peripheral monocytic cells.

Quality Segment

assay

≥98% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform/methanol (2:1) and a few drops of water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Shown to enhance spontaneous IgG, IgM, and IgA production in human peripheral monocytic cells. Note: the molecular weight is based on stearoyl-18:0.



Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Enhance spontaneous IgG, IgM, and IgA production in human peripheral monocytic cells.
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Furuse, H., et al. 1998. Exp. Brain Res. 123, 307.
Kanda, N., and Tamaki, K. 1998. J. Allergy Clin. Immunol. 102, 813.
Tsuji, S., et al. 1992. Neurochem. Int. 21, 549.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Will form a micellar solution with H₂O.
Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library