1.67585
Dip tube (PVDF) for nitric acid
pkg of 180 L (Combi-containers)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122409
NACRES:
NB.51
Recommended Products
packaging
pkg of 180 L (Combi-containers)
storage temp.
no temp limit
Related Categories
General description
in 180L Combi-containers
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service