1.50182
Hibar® RT HPLC Columns for customized packing
L × I.D. 250 mm × 4 mm, please select HPLC sorbent for Column filling
Synonym(s):
TM=["Hibar"] RT column for customized packing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115709
NACRES:
SB.52
Recommended Products
Product Name
Hibar® 250-4, suitable for HPLC
parameter
400 bar pressure
Quality Level
technique(s)
HPLC: suitable
column L × I.D.
25 cm × 4 mm
separation technique
supercritical fluid (SFC)
storage temp.
no temp limit
Related Categories
General description
HPLC column, customized packing, please specify packing material
Application
- High performance liquid chromatographic assay of amlodipine, valsartan and hydrochlorothiazide simultaneously and its application to pharmaceuticals, urine and plasma analysis: This study highlights the utilization of high-performance liquid chromatography (HPLC) for analyzing complex mixtures in pharmaceuticals and biological samples, demonstrating the sophisticated capabilities of devices like Hibar 250-4 in handling and analyzing precision samples in high-stakes environments such as pharmaceutical quality control and medical diagnostics (Shaikh JSA, Raut S, Abdul A, Pathan MAAK, 2020).
- Identification and characterization of stress degradation products of sumatriptan succinate by using LC/Q-TOF-ESI-MS/MS and NMR: The research involves advanced chromatographic techniques which could be relevant for applications using Hibar 250-4 pumps in rigorous testing environments to identify and quantify degradation products, ensuring drug safety and efficacy (Udutha S, Shankar G, Borkar RM, et al., 2018).
- Accumulation and trends in distribution of three triterpenoids in various parts of Achyranthes coynei determined using RP-UFLC analysis: This publication shows the application of ultra-fast liquid chromatography, a field where Hibar 250-4′s precision dosing could be pivotal, in the pharmacognostic study of plant materials, aiding in the rapid and effective separation of complex natural products (Upadhya V, Ankad GM, Pai SR, et al., 2014).
- Development and validation of a HPLC-PDA bioanalytical method for the simultaneous estimation of Aliskiren and Amlodipine in human plasma: This study presents a validated method for the simultaneous estimation of two pharmaceutical agents in plasma, using HPLC systems like Hibar 250-4 to achieve high precision and accuracy required for therapeutic drug monitoring and pharmacokinetic studies (Mannemala SS, Nagarajan JS, 2015).
Legal Information
Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service