1.46765
ReadyPlate 55 Slanetz&Bartley ISO 7899
for Enterococcus spp., suitable for filtration, diam. 55 mm, box of 20 plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106217
NACRES:
NA.74
Recommended Products
Agency
ISO 7899
product line
ReadyPlate™
form
plate (ready-to-use)
feature
ready-to-use
packaging
box of 20 plates
diam.
55 mm
application(s)
filtration
food and beverages
microbiology
storage temp.
15-25°C
suitability
Enterococcus spp.
Related Categories
General description
Application: Detection and enumeration of intestinal enterococci in water, Membrane filtration method
Packaging: 55 mm plate (20 plates per box)
Synonym: m-Enterococcus Agar acc. Slanetz and Bartley, Enterococcus Selective Agar
Packaging: 55 mm plate (20 plates per box)
Synonym: m-Enterococcus Agar acc. Slanetz and Bartley, Enterococcus Selective Agar
Legal Information
READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service