Skip to Content
Merck
All Photos(1)

Key Documents

1.46431

Millipore

Tryptic Soy Agar

ReadyPlate, plate diam. 90 mm, according to ISO, FDA-BAM, EP, USP

Synonym(s):

CASO agar, Casein Soybean Digest Agar, TSA, Tryptone Soya Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

EP
FDA-BAM
USP
according to ISO

Quality Level

sterility

sterile

form

plate

shelf life

4 mo.

feature

ready-to-use

packaging

pack of 100 plates
pack of 20 plates 1464310020
pack of 20 plates

manufacturer/tradename

ReadyPlate

storage condition

protect from light (Tightly closed. Dry)

plate diam.

90 mm

color

clear light yellow Media

pH

7.3 ( in H2O)

application(s)

cosmetics
environmental
food and beverages
microbiology
pharmaceutical
water monitoring

storage temp.

2-8°C

suitability

nonselective for

General description

ReadyPlate agar plates are stackable, specially designed plates that help increase safety during handling, transport, and incubation. ReadyPlate agar plates are of standard 90mm diameter. Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a medium used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation of the Tryptic Soy Agar contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids, and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, and histidine to inactivate disinfectant residues.

Application

Tryptic Soy Agar is a universal culture medium, free from indicators and inhibitors that can support the cultivation and isolation of a wide range of bacteria, yeasts, and molds in Cosmetics, Pharmaceutical, environmental, Food and Beverages samples.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The International Organization for Standardization (ISO) published the revised EN ISO 21528-1 and EN ISO 21528-2 standards in 2017.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service