Skip to Content
Merck
All Photos(1)

Key Documents

1.15045

Millipore

Meat liver agar

for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.5 (37 °C, 35 g/L in H2O)

bulk density

650 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

anaerobic bacteria

General description

for microbiology

Analysis Note

Appearance (clearness)clear to almost clearAppearance (colour)yellowish-brownpH-value (25 °C)7.4 - 7.8Growth (Clostridium perfringens ATCC 10543)good to very goodGrowth (Clostridium tetani ATCC 19406)good to very goodGrowth (Escherichia coli ATCC 25922 (WDCM 00013))good to very goodGrowth (Proteus mirabilis ATCC 14153)good to very goodGrowth (Bacteroides vulgatus ATCC 8482)fair to very goodBlacking (Clostridium perfringens ATCC 10543)+Blacking (Clostridium tetani ATCC 19406)+/-Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service