Skip to Content
Merck
All Photos(2)

Key Documents

1.11955

Sigma-Aldrich

Hemacolor® Rapid staining of blood smear

Solution 1: fixing solution

Synonym(s):

Methanol, MeOH, Hydroxymethane, Methyl alcohol, Carbinol, Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
41116121
EC Index Number:
200-659-6
NACRES:
NA.21

vapor density

1.11 (vs air)

Quality Level

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hemacolor® Rapid staining of blood smear - Solution 1: fixing solution, is a ready-to-use solution used for human-medical cell diagnosis and serves the fixing of native blood and bone-marrow smears of human origin in hematological and clinico-cytological specimen materials, for example smears of whole blood and bone marrow. The staining with Hemacolor® yields a staining result that corresponds to the Pappenheim stain, with predominantly magenta stained nuclei. This is based on the molecular interaction of the Eosin Y dye and a complex of Azur B with DNA. Both dyes assemble to an Eosin Y - Azur B-DNA complex and the intensity of the resulting stain depends on the content of Azur B and the ratio of Azur B : Eosin Y. Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. By using pH 7.2 buffered solutions with the Hemacolor® Rapid staining of blood smear solutions, a high stability of the stain and clean, precipitation-free staining results can be guaranteed.
The package is sufficient for 1250 - 5000 applications. The buffer tablets according to Weise pH 7.2 (Product number 1.09468) are also available in larger pack sizes, thus, they can be useful for higher demands. The three solutions needed for staining are all available individually as:

Hemacolor® Rapid staining of blood smear solution 1, fixing solution (Product number 1.11955)
Hemacolor® Rapid staining of blood smear solution 2, colour reagent red (Product number 1.11956)
Hemacolor® Rapid staining of blood smear solution 3, colour reagent blue (Product number 1.11957)

or they can be purchased as staining sets:

Hemacolor® Rapid staining of blood smear (Product number 1.11674) 3 x 100 ml
Hemacolor® Rapid staining of blood smear (Product number 1.11661) 3 x 500 ml

For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked.

Legal Information

HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service