Skip to Content
Merck
All Photos(1)

Key Documents

1.07381

Supelco

Lead(II) carbonate

for analysis EMSURE® ACS

Synonym(s):

Lead(II) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO3Pb
CAS Number:
Molecular Weight:
267.21
MDL number:
UNSPSC Code:
12352302
EC Index Number:
209-943-4
NACRES:
NA.21
Assay:
≥99.0% (complexometric)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

impurities

≤0.02% Substances insoluble in dilute acetic acid

pH

5.8 (20 °C, 50 g/L in H2O, slurry)

mp

315 °C (decomposition)

density

6.6 g/cm3 at 20 °C

bulk density

1300 kg/m3

anion traces

chloride (Cl-): ≤0.002%
nitrate and nitrite (as NO3-): ≤0.005%

cation traces

Ca: ≤0.01%
Cd: ≤0.002%
Cu: ≤0.002%
Fe: ≤0.002%
K: ≤0.02%
N: ≤0.02%
Na: ≤0.05%
Zn: ≤0.003%

storage temp.

2-30°C

SMILES string

[Pb+2].[O-]C(=O)[O-]

InChI

1S/CH2O3.Pb/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

MFEVGQHCNVXMER-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay (complexometric): ≥ 99.0 %
Substances insoluble in dilute acetic acid: ≤ 0.02 %
Chloride (Cl): ≤ 0.002 %
Nitrite and Nitrate (as NO₃): ≤ 0.005 %
Total nitrogen (N): ≤ 0.02 %
Ca (Calcium): ≤ 0.01 %
Cd (Cadmium): ≤ 0.002 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.02 %
Na (Sodium): ≤ 0.05 %
Zn (Zinc): ≤ 0.003 %
corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service