Skip to Content
Merck
All Photos(1)

Key Documents

T-909

Supelco

α-Hydroxytriazolam-d4 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D4H8Cl2N4O
CAS Number:
Molecular Weight:
363.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

OCC1=NN=C2N1C3=CC=C(Cl)C=C3C(C4=C([2H])C([2H])=C([2H])C([2H])=C4Cl)=NC2

InChI

1S/C17H12Cl2N4O/c18-10-5-6-14-12(7-10)17(11-3-1-2-4-13(11)19)20-8-15-21-22-16(9-24)23(14)15/h1-7,24H,8-9H2/i1D,2D,3D,4D

InChI key

BHUYWUDMVCLHND-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

a-Hydroxytriazolam is a major urinary and blood metabolite of the benzodiazepine, triazolam, a pharmaceutical used to treat severe insomnia. This stable-labeled internal standard is suitable for triazolam testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring or forensic analysis.

Application


  • Pharmacokinetic Analysis of α-Hydroxytriazolam: A study focused on the urinary excretion of α-Hydroxytriazolam following a single dose of Halcion. This research provides critical insights into the metabolism of benzodiazepines, emphasizing the role of α-Hydroxytriazolam-d₄ as a stable isotope-labeled standard in tracing metabolic pathways in clinical pharmacology (Lin et al., 2005).

  • Forensic Toxicology and Metabolite Confirmation: Another significant study utilized GC-MS to confirm the presence of urinary benzodiazepine metabolites, including α-Hydroxytriazolam. This research underscores the utility of α-Hydroxytriazolam-d₄ in forensic analysis, aiding in the detection and confirmation of benzodiazepine use and abuse in legal and medical settings (Meatherall, 1994).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service