Skip to Content
Merck
All Photos(1)

Key Documents

870312P

Avanti

16:0 DGS

Avanti Research - A Croda Brand 870312P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8
CAS Number:
Molecular Weight:
668.98
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 25 mg (870312P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870312P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(CCC(O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H72O8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(42)45-33-35(34-46-38(43)32-31-36(40)41)47-39(44)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34H2,1-2H3,(H,40,41)/t35-/m1/s1

InChI key

GNENAKXIVCYCIZ-PGUFJCEWSA-N

Application

16:0 DGS may be used in the liposomes preparation.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870312P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knut-Egil Løkling et al.
International journal of pharmaceutics, 274(1-2), 75-83 (2004-04-10)
Paramagnetic pH-responsive liposomes have recently been suggested as a promising approach for monitoring by magnetic resonance imaging (MRI) pH changes in tumours. In the present study, the effects of variations in bilayer composition on the relaxometric properties of diacylphosphatidylethanolamine (PE)/dipalmitoylglycerosuccinate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service